Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYO15B
Entrez Gene ID
80022
Gene Name
myosin XVB
Gene Aliases
MYO15BP
Chromosome Location
Chr.17: 75587545 - 75626850 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001309242.1 NP_001296171.1 14 - 15 3539 66
XM_017025120.1 XP_016880609.1 16 - 17 3246 66
XM_017025121.1 XP_016880610.1 16 - 17 3231 66
XM_017025122.1 XP_016880611.1 16 - 17 3210 66
XM_017025123.1 XP_016880612.1 16 - 17 3248 66
XM_017025125.1 XP_016880614.1 16 - 17 3245 66
XM_017025126.1 XP_016880615.1 11 - 12 2711 66
XM_017025127.1 XP_016880616.1 14 - 15 2554 66
XM_017025128.1 XP_016880617.1 15 - 16 2300 66
XM_017025129.1 XP_016880618.1 14 - 15 1532 66
XM_017025130.1 XP_016880619.1 13 - 14 1488 66
XM_017025131.1 XP_016880620.1 12 - 13 1927 66
XM_017025132.1 XP_016880621.1 11 - 12 1868 66
XM_017025133.1 XP_016880622.1 9 - 10 1393 66
XM_017025134.1 XP_016880623.1 10 - 11 1754 66
XM_017025135.1 XP_016880624.1 4 - 5 410 66
XM_017025136.1 XP_016880625.1 4 - 5 759 66
XM_017025141.1 XP_016880630.1 16 - 17 3512 66
XM_017025142.1 XP_016880631.1 16 - 17 3512 66
XM_017025144.1 XP_016880633.1 12 - 13 3522 66
GenBank mRNA AB058686.1 - 4 - 5 1646 66
AF418286.1 - 12 - 13 3772 66
AF418287.1 - 13 - 14 3860 66
AF418288.1 - 13 - 14 3572 66
AF418291.1 - 12 - 13 3522 66


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream LLGL2, scribble cell polarity complex component LLGL2 HGL, Hugl-2, LGL2 -26.2kb Sense Chr. - 75525080 - 75575209 on Build GRCh38
Downstream RecQ like helicase 5 RECQL5 RECQ5 24.3kb Anti-sense Chr. - 75626845 - 75667202 on Build GRCh38

View Full Product Details