Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MYO15B
- Entrez Gene ID
-
80022
- Gene Name
- myosin XVB
- Gene Aliases
-
MYO15BP
- Chromosome Location
-
Chr.17: 75587545 - 75626850 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BF354043.1 |
- |
1 - 2 |
|
|
Chr. 17 - 75602915 - 75603309 [-] on Build GRCh38 |
60 |
GenBankmRNA |
BX110524.1 |
- |
3 - 4 |
251 |
|
Chr. 17 - 75602510 - 75610261 [+] on Build GRCh38 |
60 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
LLGL2, scribble cell polarity complex component |
LLGL2 |
HGL, Hugl-2, LGL2 |
-27.8kb
|
Sense |
Chr. - 75525080 - 75575209 on Build GRCh38 |
Downstream |
RecQ like helicase 5 |
RECQL5 |
RECQ5 |
23.5kb
|
Anti-sense |
Chr. - 75626845 - 75667202 on Build GRCh38 |
View Full Product Details