Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REV1
Entrez Gene ID
51455
Gene Name
REV1, DNA directed polymerase
Gene Aliases
AIBP80, REV1L
Chromosome Location
Chr.2: 99400475 - 99490123 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037872.2 NP_001032961.1 12 - 13 2203 99
NM_001321454.1 NP_001308383.1 13 - 14 2314 99
NM_001321455.1 NP_001308384.1 13 - 14 2360 99
NM_001321458.1 NP_001308387.1 13 - 14 2330 99
NM_001321459.1 NP_001308388.1 13 - 14 2406 99
NM_001321460.1 NP_001308389.1 13 - 14 2403 99
NM_016316.3 NP_057400.1 12 - 13 2206 99
NR_135649.1 13 - 14 2403 99
NR_135650.1 13 - 14 2360 99
NR_135651.1 12 - 13 2203 99
NR_135652.1 12 - 13 2206 99
NR_135653.1 12 - 13 2206 99
XM_011511341.2 XP_011509643.1 6 - 7 823 99
XM_017004309.1 XP_016859798.1 13 - 14 2727 99
XM_017004310.1 XP_016859799.1 13 - 14 2093 99
XM_017004311.1 XP_016859800.1 13 - 14 2093 99
XM_017004312.1 XP_016859801.1 13 - 14 2306 99
XM_017004313.1 XP_016859802.1 12 - 13 3932 99
XM_017004314.1 XP_016859803.1 9 - 10 1583 99
XM_017004315.1 XP_016859804.1 9 - 10 1567 99
GenBank mRNA AB047646.1 - 12 - 13 2128 99
AF151538.1 - 12 - 13 2159 99
AF206019.1 - 12 - 13 2168 99
AF357886.1 - 12 - 13 2124 99
AJ131720.1 - 4 - 5 525 99
AK025176.1 - 4 - 5 485 99
AK307719.1 - 12 - 13 2117 99
BC037734.1 - 13 - 14 2341 99
BC130411.1 - 11 - 12 1974 99

View Full Product Details