The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037872.2 | NP_001032961.1 | 7 - 8 | 1571 | 124 |
NM_001321454.1 | NP_001308383.1 | 7 - 8 | 1571 | 124 | |
NM_001321455.1 | NP_001308384.1 | 8 - 9 | 1728 | 124 | |
NM_001321458.1 | NP_001308387.1 | 7 - 8 | 1571 | 124 | |
NM_001321459.1 | NP_001308388.1 | 7 - 8 | 1571 | 124 | |
NM_001321460.1 | NP_001308389.1 | 7 - 8 | 1571 | 124 | |
NM_016316.3 | NP_057400.1 | 7 - 8 | 1571 | 124 | |
NR_135649.1 | 7 - 8 | 1571 | 124 | ||
NR_135650.1 | 8 - 9 | 1728 | 124 | ||
NR_135651.1 | 7 - 8 | 1571 | 124 | ||
NR_135652.1 | 7 - 8 | 1571 | 124 | ||
NR_135653.1 | 7 - 8 | 1571 | 124 | ||
XM_017004309.1 | XP_016859798.1 | 7 - 8 | 1984 | 124 | |
XM_017004310.1 | XP_016859799.1 | 7 - 8 | 1350 | 124 | |
XM_017004311.1 | XP_016859800.1 | 7 - 8 | 1350 | 124 | |
XM_017004312.1 | XP_016859801.1 | 7 - 8 | 1566 | 124 | |
XM_017004313.1 | XP_016859802.1 | 7 - 8 | 3300 | 124 | |
XM_017004314.1 | XP_016859803.1 | 2 - 3 | 624 | 124 | |
XM_017004315.1 | XP_016859804.1 | 2 - 3 | 624 | 124 | |
GenBank mRNA | AB047646.1 | - | 7 - 8 | 1496 | 124 |
AF151538.1 | - | 7 - 8 | 1524 | 124 | |
AF206019.1 | - | 7 - 8 | 1533 | 124 | |
AF357886.1 | - | 7 - 8 | 1489 | 124 | |
AK307719.1 | - | 7 - 8 | 1482 | 124 | |
BC037734.1 | - | 8 - 9 | 1709 | 124 | |
BC130411.1 | - | 6 - 7 | 1342 | 124 | |
BP269179.1 | - | 2 - 3 | 325 | 124 | |
DA304601.1 | - | 2 - 3 | 235 | 124 |