The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015904.3 | NP_056988.3 | 12 - 13 | 2248 | 119 |
GenBank mRNA | AB018284.1 | - | 12 - 13 | 2214 | 119 |
AF078035.1 | - | 12 - 13 | 2181 | 119 | |
AJ006776.1 | - | 12 - 13 | 2204 | 119 | |
AK001386.1 | - | 3 - 4 | 119 | ||
AK308176.1 | - | 12 - 13 | 2230 | 119 | |
AL133563.1 | - | 9 - 10 | 1800 | 119 | |
BC032639.1 | - | 12 - 13 | 2170 | 119 |