The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015904.3 | NP_056988.3 | 2 - 3 | 341 | 108 |
GenBank mRNA | AF078035.1 | - | 2 - 3 | 274 | 108 |
AJ006776.1 | - | 2 - 3 | 297 | 108 | |
AK308175.1 | - | 2 - 3 | 323 | 108 | |
AK308176.1 | - | 2 - 3 | 323 | 108 | |
BC006970.1 | - | 2 - 3 | 191 | 108 | |
BC022260.1 | - | 2 - 3 | 219 | 108 | |
BC032639.1 | - | 2 - 3 | 263 | 108 | |
BX485839.1 | - | 2 - 3 | 341 | 108 |