Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VIT
Entrez Gene ID
5212
Gene Name
vitrin
Gene Aliases
-
Chromosome Location
Chr.2: 36696685 - 36814794 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177969.1 NP_001171440.1 4 - 5 571 78
NM_001177970.1 NP_001171441.1 4 - 5 571 78
NM_001177971.1 NP_001171442.1 4 - 5 571 78
NM_001177972.1 NP_001171443.1 4 - 5 571 78
NM_053276.3 NP_444506.2 4 - 5 571 78
XM_011532905.1 XP_011531207.1 4 - 5 468 78
XM_011532906.1 XP_011531208.1 4 - 5 468 78
XM_017004325.1 XP_016859814.1 4 - 5 576 78
XM_017004326.1 XP_016859815.1 4 - 5 468 78
XM_017004327.1 XP_016859816.1 2 - 3 474 78
GenBank mRNA AK056772.1 - 4 - 5 490 78
AK292149.1 - 4 - 5 468 78
AK299426.1 - 4 - 5 554 78
AL832679.1 - 4 - 5 414 78
AY358338.1 - 4 - 5 456 78
BC061519.1 - 4 - 5 497 78
BC128260.1 - 4 - 5 384 78

View Full Product Details