The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177969.1 | NP_001171440.1 | 6 - 7 | 788 | 99 |
NM_001177970.1 | NP_001171441.1 | 6 - 7 | 788 | 99 | |
NM_001177971.1 | NP_001171442.1 | 6 - 7 | 788 | 99 | |
NM_053276.3 | NP_444506.2 | 6 - 7 | 788 | 99 | |
XM_017004325.1 | XP_016859814.1 | 6 - 7 | 793 | 99 | |
XM_017004327.1 | XP_016859816.1 | 4 - 5 | 691 | 99 | |
GenBank mRNA | AF063833.1 | - | 6 - 7 | 612 | 99 |
AK056772.1 | - | 6 - 7 | 707 | 99 | |
AY358338.1 | - | 6 - 7 | 673 | 99 | |
BC128260.1 | - | 6 - 7 | 601 | 99 |