Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC43A2
Entrez Gene ID
124935
Gene Name
solute carrier family 43 member 2
Gene Aliases
LAT4
Chromosome Location
Chr.17: 1569254 - 1630014 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017024179.1 XP_016879668.1 1 - 2 410 88

View Full Product Details