The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012401.3 | NP_036533.2 | 23 - 24 | 4087 | 79 |
XM_005261909.1 | XP_005261966.1 | 24 - 25 | 4135 | 79 | |
XM_005261910.2 | XP_005261967.1 | 24 - 25 | 4146 | 79 | |
XM_005261911.4 | XP_005261968.1 | 23 - 24 | 4085 | 79 | |
XM_006724413.2 | XP_006724476.1 | 25 - 26 | 4205 | 79 | |
XM_011530682.2 | XP_011528984.1 | 24 - 25 | 4286 | 79 | |
XM_017028703.1 | XP_016884192.1 | 25 - 26 | 4186 | 79 | |
XM_017028704.1 | XP_016884193.1 | 24 - 25 | 4139 | 79 | |
GenBank mRNA | AB002313.1 | - | 22 - 23 | 4004 | 79 |
AB075511.1 | - | 8 - 9 | 1197 | 79 | |
AK025701.1 | - | 10 - 11 | 1455 | 79 | |
AK056543.1 | - | 6 - 7 | 1256 | 79 | |
AK074932.1 | - | 5 - 6 | 723 | 79 | |
AK126394.1 | - | 4 - 5 | 796 | 79 | |
BC148225.1 | - | 1 - 2 | 266 | 79 | |
BC148280.1 | - | 1 - 2 | 266 | 79 | |
S76730.1 | - | 1 - 2 | 239 | 79 |