The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173050.3 | NP_766638.2 | 17 - 18 | 2355 | 74 |
XM_005261752.1 | XP_005261809.1 | 18 - 19 | 2406 | 74 | |
XM_011530389.2 | XP_011528691.1 | 18 - 19 | 2400 | 74 | |
XM_011530390.2 | XP_011528692.1 | 17 - 18 | 2285 | 74 | |
GenBank mRNA | AF525689.1 | - | 18 - 19 | 2248 | 74 |
AK127719.1 | - | 14 - 15 | 1828 | 74 | |
BX280767.1 | - | 2 - 3 | 333 | 74 |