The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199096.1 | NP_001186025.1 | 12 - 13 | 1265 | 97 |
NM_001199097.1 | NP_001186026.1 | 13 - 14 | 1315 | 97 | |
NM_001199098.1 | NP_001186027.1 | 13 - 14 | 1246 | 97 | |
NM_001199099.1 | NP_001186028.1 | 13 - 14 | 1231 | 97 | |
NM_001286464.1 | NP_001273393.1 | 13 - 14 | 1366 | 97 | |
NM_003933.4 | NP_003924.2 | 13 - 14 | 1423 | 97 | |
XM_011522728.1 | XP_011521030.1 | 13 - 14 | 1969 | 97 | |
XM_011522729.1 | XP_011521031.1 | 13 - 14 | 1969 | 97 | |
XM_011522730.2 | XP_011521032.1 | 13 - 14 | 1969 | 97 | |
GenBank mRNA | AB018277.2 | - | 13 - 14 | 1275 | 97 |
AK294576.1 | - | 2 - 3 | 186 | 97 | |
AK294744.1 | - | 13 - 14 | 1191 | 97 | |
AK295645.1 | - | 13 - 14 | 1205 | 97 | |
AK299309.1 | - | 13 - 14 | 1275 | 97 | |
AK315059.1 | - | 13 - 14 | 1423 | 97 | |
AL834321.1 | - | 13 - 14 | 1317 | 97 | |
BC104966.1 | - | 13 - 14 | 1286 | 97 | |
BC112129.1 | - | 13 - 14 | 1298 | 97 |