The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020730.2 | NP_065781.1 | 1 - 2 | 455 | 159 |
XM_017029322.1 | XP_016884811.1 | 1 - 2 | 444 | 159 | |
XM_017029323.1 | XP_016884812.1 | 1 - 2 | 444 | 159 | |
XM_017029324.1 | XP_016884813.1 | 1 - 2 | 464 | 159 | |
GenBank mRNA | AB033058.1 | - | 1 - 2 | 453 | 159 |
AK304020.1 | - | 1 - 2 | 168 | 159 | |
AK316518.1 | - | 1 - 2 | 154 | 159 | |
CN357435.1 | - | 1 - 2 | 450 | 159 |