Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RADIL
Entrez Gene ID
55698
Gene Name
Rap associating with DIL domain
Gene Aliases
RASIP2
Chromosome Location
Chr.7: 4799109 - 4883704 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018059.4 NP_060529.4 4 - 5 1590 69
GenBank mRNA AB058752.1 - 4 - 5 1534 69
AK001186.1 - 1 - 2 573 69
AK294745.1 - 4 - 5 1550 69
AK296325.1 - 3 - 4 1111 69
AK302549.1 - - 1582 69
BC117317.1 - 3 - 4 1476 69
BC126311.1 - 3 - 4 1476 69
BC143526.1 - 3 - 4 1476 69
EF560723.1 - 3 - 4 1499 69

View Full Product Details