Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSC2
Entrez Gene ID
7249
Gene Name
tuberous sclerosis 2
Gene Aliases
LAM, PPP1R160, TSC4
Chromosome Location
Chr.16: 2047804 - 2088720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318827.1 NP_001305756.1 24 - 25 2922 82
NM_001318829.1 NP_001305758.1 24 - 25 2866 82
XM_005255529.4 XP_005255586.2 25 - 26 2920 82
XM_005255531.4 XP_005255588.2 25 - 26 2920 82
XM_011522639.2 XP_011520941.1 25 - 26 2920 82
XM_017023617.1 XP_016879106.1 24 - 25 3013 82
GenBank mRNA AK295672.1 - 24 - 25 2753 82
KJ534954.1 - 24 - 25 2833 82
KJ535091.1 - 24 - 25 2833 82

View Full Product Details