Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS17
Entrez Gene ID
170691
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 17
Gene Aliases
-
Chromosome Location
Chr.15: 99971438 - 100342924 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139057.3 NP_620688.2 10 - 11 1576 89
XM_005254872.3 XP_005254929.1 10 - 11 2492 89
XM_011521312.2 XP_011519614.1 10 - 11 2492 89
XM_017021973.1 XP_016877462.1 11 - 12 2625 89
XM_017021974.1 XP_016877463.1 11 - 12 2627 89
XM_017021975.1 XP_016877464.1 11 - 12 2625 89
XM_017021976.1 XP_016877465.1 10 - 11 1337 89
XM_017021977.1 XP_016877466.1 11 - 12 2624 89
XM_017021978.1 XP_016877467.1 4 - 5 3883 89
XM_017021979.1 XP_016877468.1 3 - 4 333 89
XM_017021980.1 XP_016877469.1 3 - 4 543 89
XM_017021981.1 XP_016877470.1 11 - 12 2622 89
XM_017021984.1 XP_016877473.1 9 - 10 1151 89
GenBank mRNA AJ315735.1 - 10 - 11 1493 89
AK131344.1 - 9 - 10 1122 89

View Full Product Details