Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USP53
Entrez Gene ID
54532
Gene Name
ubiquitin specific peptidase 53
Gene Aliases
-
Chromosome Location
Chr.4: 119212583 - 119295518 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019050.2 NP_061923.2 11 - 12 2033 99
XM_005263073.3 XP_005263130.1 10 - 11 1907 99
XM_005263076.2 XP_005263133.1 11 - 12 2072 99
XM_005263077.3 XP_005263134.1 11 - 12 2072 99
XM_006714240.1 XP_006714303.1 9 - 10 1108 99
XM_011532037.2 XP_011530339.1 11 - 12 2085 99
XM_011532038.2 XP_011530340.1 10 - 11 1362 99
XM_017008312.1 XP_016863801.1 12 - 13 2199 99
XM_017008313.1 XP_016863802.1 11 - 12 1884 99
XM_017008314.1 XP_016863803.1 11 - 12 1883 99
XM_017008315.1 XP_016863804.1 11 - 12 1953 99
XM_017008316.1 XP_016863805.1 10 - 11 1924 99
XM_017008317.1 XP_016863806.1 10 - 11 1896 99
XM_017008318.1 XP_016863807.1 11 - 12 2018 99
GenBank mRNA AB037771.1 - 4 - 5 482 99
AJ583824.1 - 8 - 9 964 99
CR749490.1 - 12 - 13 2148 99

View Full Product Details