The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173482.1 | NP_001166953.1 | 4 - 5 | 554 | 91 |
NM_016302.3 | NP_057386.2 | 4 - 5 | 557 | 91 | |
XM_005265202.3 | XP_005265259.1 | 3 - 4 | 438 | 91 | |
XM_011533791.2 | XP_011532093.1 | 4 - 5 | 544 | 91 | |
GenBank mRNA | AF117230.1 | - | 4 - 5 | 521 | 91 |
AK027507.1 | - | 4 - 5 | 535 | 91 | |
AK058191.1 | - | 4 - 5 | 531 | 91 | |
AK312577.1 | - | 4 - 5 | 535 | 91 | |
BC017419.1 | - | 4 - 5 | 545 | 91 | |
BC067811.1 | - | 4 - 5 | 538 | 91 | |
CR627060.1 | - | 3 - 4 | 424 | 91 |