Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCAF8
Entrez Gene ID
50717
Gene Name
DDB1 and CUL4 associated factor 8
Gene Aliases
GAN2, H326, WDR42A
Chromosome Location
Chr.1: 160215715 - 160262560 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BP359560.1 - 1 - 2 210 97

View Full Product Details