The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015726.3 | NP_056541.2 | 2 - 3 | 188 | 75 |
NR_028103.1 | 2 - 3 | 488 | 75 | ||
NR_028105.1 | 2 - 3 | 488 | 75 | ||
NR_028106.1 | 2 - 3 | 255 | 75 | ||
GenBank mRNA | AK026481.1 | - | 2 - 3 | 252 | 75 |
AK223475.1 | - | 1 - 2 | 43 | 75 | |
AK303188.1 | - | 2 - 3 | 113 | 75 | |
BC013107.1 | - | 2 - 3 | 204 | 75 | |
BC098271.1 | - | 2 - 3 | 94 | 75 | |
BC099709.1 | - | 2 - 3 | 94 | 75 | |
BC099846.1 | - | 2 - 3 | 94 | 75 | |
BC111063.1 | - | 2 - 3 | 385 | 75 | |
CR749801.1 | - | 2 - 3 | 411 | 75 |