The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323041.1 | NP_001309970.1 | 6 - 7 | 1149 | 106 |
NM_001323042.1 | NP_001309971.1 | 6 - 7 | 633 | 106 | |
NM_001323043.1 | NP_001309972.1 | 6 - 7 | 1034 | 106 | |
NM_001323044.1 | NP_001309973.1 | 6 - 7 | 1407 | 106 | |
NM_001323045.1 | NP_001309974.1 | 7 - 8 | 1118 | 106 | |
NM_001323046.1 | NP_001309975.1 | 4 - 5 | 805 | 106 | |
NM_001323047.1 | NP_001309976.1 | 6 - 7 | 1149 | 106 | |
NM_001323048.1 | NP_001309977.1 | 6 - 7 | 1034 | 106 | |
NM_001323049.1 | NP_001309978.1 | 5 - 6 | 1323 | 106 | |
NM_001323050.1 | NP_001309979.1 | 7 - 8 | 1118 | 106 | |
NM_001323051.1 | NP_001309980.1 | 6 - 7 | 1407 | 106 | |
NM_001323052.1 | NP_001309981.1 | 6 - 7 | 1034 | 106 | |
NM_001323053.1 | NP_001309982.1 | 6 - 7 | 1149 | 106 | |
NM_006608.2 | NP_006599.2 | 5 - 6 | 938 | 106 | |
NR_136564.1 | 7 - 8 | 1233 | 106 | ||
NR_136565.1 | 6 - 7 | 1034 | 106 | ||
NR_136566.1 | 6 - 7 | 1149 | 106 | ||
NR_136567.1 | 6 - 7 | 1034 | 106 | ||
NR_136568.1 | 7 - 8 | 1118 | 106 | ||
GenBank mRNA | AJ011863.1 | - | 5 - 6 | 517 | 106 |
AK294749.1 | - | 5 - 6 | 664 | 106 | |
BC002447.2 | - | 6 - 7 | 1082 | 106 |