Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYL3
Entrez Gene ID
4634
Gene Name
myosin light chain 3
Gene Aliases
CMH8, MLC-lV/sb, MLC1SB, MLC1V, VLC1, VLCl
Chromosome Location
Chr.3: 46857867 - 46863483 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000258.2 NP_000249.1 2 - 3 243 82
GenBank mRNA AF174483.1 - 2 - 3 150 82
AK312044.1 - 2 - 3 204 82
BC009790.1 - 2 - 3 208 82
BP316922.1 - 2 - 3 243 82
M24122.1 - 2 - 3 200 82
X07373.1 - 2 - 3 150 82

View Full Product Details