The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010898.2 | NP_001010898.1 | 7 - 8 | 1563 | 71 |
XM_006710643.2 | XP_006710706.1 | 7 - 8 | 1271 | 71 | |
GenBank mRNA | AK289982.1 | - | 7 - 8 | 1559 | 71 |
BC140908.1 | - | 6 - 7 | 1148 | 71 |