The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024713.2 | NP_078989.1 | 6 - 7 | 760 | 85 |
XM_005254668.3 | XP_005254725.1 | 6 - 7 | 810 | 85 | |
XM_017022571.1 | XP_016878060.1 | 7 - 8 | 844 | 85 | |
XM_017022572.1 | XP_016878061.1 | 7 - 8 | 1149 | 85 | |
XM_017022573.1 | XP_016878062.1 | 6 - 7 | 970 | 85 | |
XM_017022574.1 | XP_016878063.1 | 7 - 8 | 823 | 85 | |
GenBank mRNA | AK293021.1 | - | 6 - 7 | 697 | 85 |
AK293695.1 | - | 5 - 6 | 629 | 85 | |
AK311338.1 | - | 5 - 6 | 480 | 85 | |
AL136908.1 | - | 6 - 7 | 686 | 85 | |
AL833631.1 | - | 6 - 7 | 744 | 85 | |
BC026234.1 | - | 6 - 7 | 652 | 85 | |
BC111000.1 | - | 6 - 7 | 721 | 85 |