Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANO2
Entrez Gene ID
57101
Gene Name
anoctamin 2
Gene Aliases
C12orf3, TMEM16B
Chromosome Location
Chr.12: 5560241 - 5948790 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278596.1 NP_001265525.1 25 - 26 2678 59
NM_001278597.1 NP_001265526.1 25 - 26 2696 59
XM_006718988.3 XP_006719051.1 23 - 24 2953 59
XM_011520975.2 XP_011519277.1 23 - 24 5239 59
XM_011520978.2 XP_011519280.1 22 - 23 2551 59
XM_017019672.1 XP_016875161.1 23 - 24 2953 59
GenBank mRNA AJ272204.1 - 25 - 26 2696 59
AK024010.1 - 8 - 9 960 59
BM710588.1 - 1 - 2 110 59
FJ384095.1 - 25 - 26 2652 59

View Full Product Details