Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANO2
Entrez Gene ID
57101
Gene Name
anoctamin 2
Gene Aliases
C12orf3, TMEM16B
Chromosome Location
Chr.12: 5560241 - 5948790 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278596.1 NP_001265525.1 26 - 27 2785 109
NM_001278597.1 NP_001265526.1 26 - 27 2803 109
XM_006718988.3 XP_006719051.1 24 - 25 3060 109
XM_011520975.2 XP_011519277.1 24 - 25 5346 109
XM_011520978.2 XP_011519280.1 23 - 24 2658 109
GenBank mRNA AJ272204.1 - 26 - 27 2803 109
AK024010.1 - 9 - 10 1067 109
BM710588.1 - 2 - 3 217 109
FJ384095.1 - 26 - 27 2759 109

View Full Product Details