The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006718988.3 | XP_006719051.1 | 4 - 5 | 967 | 90 |
XM_011520975.2 | XP_011519277.1 | 4 - 5 | 3253 | 90 | |
XM_011520978.2 | XP_011519280.1 | 3 - 4 | 565 | 90 | |
XM_017019672.1 | XP_016875161.1 | 4 - 5 | 967 | 90 | |
GenBank mRNA | AB593159.1 | - | 4 - 5 | 677 | 90 |
DB071682.1 | - | 2 - 3 | 303 | 90 |