Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANO2
Entrez Gene ID
57101
Gene Name
anoctamin 2
Gene Aliases
C12orf3, TMEM16B
Chromosome Location
Chr.12: 5560241 - 5948790 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278596.1 NP_001265525.1 10 - 11 1039 92
NM_001278597.1 NP_001265526.1 10 - 11 1057 92
XM_006718988.3 XP_006719051.1 9 - 10 1326 92
XM_011520975.2 XP_011519277.1 9 - 10 3612 92
XM_011520978.2 XP_011519280.1 8 - 9 924 92
XM_017019672.1 XP_016875161.1 9 - 10 1326 92
GenBank mRNA AJ272204.1 - 10 - 11 1057 92
FJ384095.1 - 10 - 11 1013 92

View Full Product Details