The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031715.2 | NP_001026885.1 | 2 - 3 | 272 | 142 |
NM_022784.2 | NP_073621.2 | 2 - 3 | 272 | 142 | |
XM_011521907.2 | XP_011520209.1 | 2 - 3 | 296 | 142 | |
XM_011521912.2 | XP_011520214.1 | 2 - 3 | 296 | 142 | |
XM_011521913.2 | XP_011520215.1 | 2 - 3 | 297 | 142 | |
XM_011521915.2 | XP_011520217.1 | 2 - 3 | 296 | 142 | |
XM_011521917.2 | XP_011520219.1 | 1 - 2 | 124 | 142 | |
XM_011521922.2 | XP_011520224.1 | 2 - 3 | 299 | 142 | |
XM_011521923.2 | XP_011520225.1 | 2 - 3 | 300 | 142 | |
XM_017022491.1 | XP_016877980.1 | 2 - 3 | 296 | 142 | |
XM_017022492.1 | XP_016877981.1 | 2 - 3 | 296 | 142 | |
XM_017022493.1 | XP_016877982.1 | 2 - 3 | 296 | 142 | |
XM_017022494.1 | XP_016877983.1 | 2 - 3 | 296 | 142 | |
XM_017022495.1 | XP_016877984.1 | 2 - 3 | 296 | 142 | |
XM_017022496.1 | XP_016877985.1 | 2 - 3 | 296 | 142 | |
XM_017022497.1 | XP_016877986.1 | 2 - 3 | 296 | 142 | |
XM_017022498.1 | XP_016877987.1 | 2 - 3 | 296 | 142 | |
XM_017022499.1 | XP_016877988.1 | 2 - 3 | 296 | 142 | |
GenBank mRNA | AK292478.1 | - | 2 - 3 | 224 | 142 |
AK304303.1 | - | 2 - 3 | 224 | 142 | |
AY014282.2 | - | 2 - 3 | 243 | 142 | |
BC048964.1 | - | 2 - 3 | 222 | 142 |