Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IQCH
Entrez Gene ID
64799
Gene Name
IQ motif containing H
Gene Aliases
NYDSP5
Chromosome Location
Chr.15: 67254772 - 67501804 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031715.2 NP_001026885.1 3 - 4 359 140
NM_022784.2 NP_073621.2 3 - 4 359 140
XM_011521907.2 XP_011520209.1 3 - 4 383 140
XM_011521912.2 XP_011520214.1 3 - 4 383 140
XM_011521913.2 XP_011520215.1 3 - 4 384 140
XM_011521915.2 XP_011520217.1 3 - 4 383 140
XM_011521917.2 XP_011520219.1 2 - 3 211 140
XM_011521922.2 XP_011520224.1 3 - 4 386 140
XM_011521923.2 XP_011520225.1 3 - 4 387 140
XM_017022491.1 XP_016877980.1 3 - 4 383 140
XM_017022492.1 XP_016877981.1 3 - 4 383 140
XM_017022493.1 XP_016877982.1 3 - 4 383 140
XM_017022494.1 XP_016877983.1 3 - 4 383 140
XM_017022495.1 XP_016877984.1 3 - 4 383 140
XM_017022496.1 XP_016877985.1 3 - 4 383 140
XM_017022497.1 XP_016877986.1 3 - 4 383 140
XM_017022498.1 XP_016877987.1 3 - 4 383 140
XM_017022499.1 XP_016877988.1 3 - 4 383 140
GenBank mRNA AK292478.1 - 3 - 4 311 140
AK304303.1 - 3 - 4 311 140
AY014282.2 - 3 - 4 330 140
BC048964.1 - 3 - 4 309 140

View Full Product Details