Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IDH3G
Entrez Gene ID
3421
Gene Name
isocitrate dehydrogenase 3 (NAD(+)) gamma
Gene Aliases
H-IDHG
Chromosome Location
Chr.X: 153785766 - 153794523 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004135.3 NP_004126.1 6 - 7 605 66
NM_174869.2 NP_777358.1 6 - 7 605 66
GenBank mRNA AB209206.1 - 1 - 2 3216 66
BC000933.2 - 6 - 7 451 66
BC001902.1 - 6 - 7 446 66
BI831096.1 - 6 - 7 603 66
U40272.1 - 6 - 7 600 66
U69268.1 - 2 - 3 156 66
Z68907.1 - 6 - 7 487 66

View Full Product Details