The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320941.1 | NP_001307870.1 | 10 - 11 | 1505 | 130 |
NM_001320942.1 | NP_001307871.1 | 11 - 12 | 1829 | 130 | |
NM_004136.3 | NP_004127.1 | 11 - 12 | 1575 | 130 | |
GenBank mRNA | AB209403.1 | - | 11 - 12 | 1509 | 130 |
AK027033.1 | - | 10 - 11 | 1339 | 130 | |
AK027034.1 | - | 2 - 3 | 193 | 130 | |
AK292992.1 | - | 11 - 12 | 1459 | 130 | |
BC117481.1 | - | 11 - 12 | 1442 | 130 | |
BC117483.1 | - | 11 - 12 | 1442 | 130 | |
BC143948.1 | - | 10 - 11 | 1372 | 130 | |
M58511.1 | - | 11 - 12 | 1413 | 130 | |
U20180.1 | - | 10 - 11 | 1380 | 130 |