The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025258.2 | NP_079534.2 | 10 - 11 | 1740 | 79 |
XM_005249427.2 | XP_005249484.1 | 10 - 11 | 1740 | 79 | |
XM_017011327.1 | XP_016866816.1 | 9 - 10 | 1775 | 79 | |
XM_017011328.1 | XP_016866817.1 | 10 - 11 | 1886 | 79 | |
XM_017011329.1 | XP_016866818.1 | 8 - 9 | 1284 | 79 | |
GenBank mRNA | AJ245418.1 | - | 5 - 6 | 610 | 79 |
AK304266.1 | - | 9 - 10 | 1513 | 79 | |
BC114625.1 | - | 9 - 10 | 1499 | 79 |