The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025258.2 | NP_079534.2 | 4 - 5 | 854 | 89 |
XM_005249427.2 | XP_005249484.1 | 4 - 5 | 854 | 89 | |
XM_017011327.1 | XP_016866816.1 | 3 - 4 | 889 | 89 | |
XM_017011328.1 | XP_016866817.1 | 4 - 5 | 1000 | 89 | |
XM_017011329.1 | XP_016866818.1 | 2 - 3 | 379 | 89 | |
GenBank mRNA | AK304058.1 | - | 4 - 5 | 830 | 89 |
AK304266.1 | - | 3 - 4 | 627 | 89 | |
BC114625.1 | - | 3 - 4 | 613 | 89 |