The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003253.2 | NP_003244.2 | 12 - 13 | 2854 | 79 |
XM_005261037.1 | XP_005261094.1 | 11 - 12 | 2987 | 79 | |
XM_005261038.2 | XP_005261095.1 | 10 - 11 | 2721 | 79 | |
XM_005261040.2 | XP_005261097.1 | 11 - 12 | 2987 | 79 | |
XM_011529711.1 | XP_011528013.1 | 12 - 13 | 2938 | 79 | |
XM_011529712.1 | XP_011528014.1 | 12 - 13 | 2862 | 79 | |
XM_011529713.1 | XP_011528015.1 | 12 - 13 | 3272 | 79 | |
XM_017028447.1 | XP_016883936.1 | 12 - 13 | 3041 | 79 | |
XM_017028448.1 | XP_016883937.1 | 12 - 13 | 3011 | 79 | |
XM_017028449.1 | XP_016883938.1 | 11 - 12 | 2918 | 79 | |
XM_017028450.1 | XP_016883939.1 | 11 - 12 | 2938 | 79 | |
XM_017028451.1 | XP_016883940.1 | 11 - 12 | 3418 | 79 | |
XM_017028452.1 | XP_016883941.1 | 13 - 14 | 3218 | 79 | |
XM_017028453.1 | XP_016883942.1 | 11 - 12 | 2893 | 79 | |
GenBank mRNA | AB209101.1 | - | 8 - 9 | 1870 | 79 |
BC117192.1 | - | 9 - 10 | 2429 | 79 | |
BC117196.1 | - | 9 - 10 | 2429 | 79 | |
BC143970.1 | - | 9 - 10 | 2429 | 79 | |
U16296.1 | - | 12 - 13 | 2855 | 79 |