The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003253.2 | NP_003244.2 | 13 - 14 | 2956 | 153 |
XM_005261037.1 | XP_005261094.1 | 12 - 13 | 3089 | 153 | |
XM_005261038.2 | XP_005261095.1 | 11 - 12 | 2823 | 153 | |
XM_005261040.2 | XP_005261097.1 | 12 - 13 | 3089 | 153 | |
XM_011529711.1 | XP_011528013.1 | 13 - 14 | 3040 | 153 | |
XM_011529712.1 | XP_011528014.1 | 13 - 14 | 2964 | 153 | |
XM_011529713.1 | XP_011528015.1 | 13 - 14 | 3374 | 153 | |
XM_017028447.1 | XP_016883936.1 | 13 - 14 | 3143 | 153 | |
XM_017028448.1 | XP_016883937.1 | 13 - 14 | 3113 | 153 | |
XM_017028449.1 | XP_016883938.1 | 12 - 13 | 3020 | 153 | |
XM_017028450.1 | XP_016883939.1 | 12 - 13 | 3040 | 153 | |
XM_017028451.1 | XP_016883940.1 | 12 - 13 | 3520 | 153 | |
XM_017028452.1 | XP_016883941.1 | 14 - 15 | 3320 | 153 | |
XM_017028453.1 | XP_016883942.1 | 12 - 13 | 2995 | 153 | |
GenBank mRNA | AB209101.1 | - | 9 - 10 | 1972 | 153 |
BC117192.1 | - | 10 - 11 | 2531 | 153 | |
BC117196.1 | - | 10 - 11 | 2531 | 153 | |
BC143970.1 | - | 10 - 11 | 2531 | 153 | |
U16296.1 | - | 13 - 14 | 2957 | 153 |