The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003253.2 | NP_003244.2 | 5 - 6 | 1426 | 58 |
XM_005261037.1 | XP_005261094.1 | 4 - 5 | 1559 | 58 | |
XM_005261038.2 | XP_005261095.1 | 3 - 4 | 1293 | 58 | |
XM_005261040.2 | XP_005261097.1 | 4 - 5 | 1559 | 58 | |
XM_011529711.1 | XP_011528013.1 | 5 - 6 | 1510 | 58 | |
XM_011529712.1 | XP_011528014.1 | 5 - 6 | 1434 | 58 | |
XM_011529713.1 | XP_011528015.1 | 5 - 6 | 1844 | 58 | |
XM_017028447.1 | XP_016883936.1 | 5 - 6 | 1613 | 58 | |
XM_017028448.1 | XP_016883937.1 | 5 - 6 | 1583 | 58 | |
XM_017028449.1 | XP_016883938.1 | 4 - 5 | 1490 | 58 | |
XM_017028450.1 | XP_016883939.1 | 4 - 5 | 1510 | 58 | |
XM_017028451.1 | XP_016883940.1 | 4 - 5 | 1990 | 58 | |
XM_017028452.1 | XP_016883941.1 | 6 - 7 | 1790 | 58 | |
XM_017028453.1 | XP_016883942.1 | 4 - 5 | 1465 | 58 | |
GenBank mRNA | AB209101.1 | - | 1 - 2 | 442 | 58 |
BC117192.1 | - | 2 - 3 | 1001 | 58 | |
BC117196.1 | - | 2 - 3 | 1001 | 58 | |
BC143970.1 | - | 2 - 3 | 1001 | 58 | |
BC143980.1 | - | 2 - 3 | 1001 | 58 | |
U16296.1 | - | 5 - 6 | 1427 | 58 |