The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001197297.1 | NP_001184226.1 | 2 - 3 | 220 | 109 |
NM_002040.3 | NP_002031.2 | 2 - 3 | 657 | 109 | |
XM_005260938.4 | XP_005260995.1 | 2 - 3 | 292 | 109 | |
XM_011529520.2 | XP_011527822.1 | 3 - 4 | 696 | 109 | |
XM_011529521.2 | XP_011527823.1 | 4 - 5 | 787 | 109 | |
XM_017028313.1 | XP_016883802.1 | 2 - 3 | 303 | 109 | |
GenBank mRNA | AK223164.1 | - | 2 - 3 | 259 | 109 |
AK313545.1 | - | 2 - 3 | 454 | 109 | |
BC035031.2 | - | 2 - 3 | 290 | 109 | |
BX647755.1 | - | 2 - 3 | 598 | 109 | |
D13318.1 | - | 2 - 3 | 262 | 109 | |
EU159453.1 | - | 1 - 2 | 79 | 109 | |
JQ313900.1 | - | 1 - 2 | 70 | 109 | |
U13044.1 | - | 2 - 3 | 255 | 109 |