The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001197297.1 | NP_001184226.1 | 7 - 8 | 956 | 89 |
NM_002040.3 | NP_002031.2 | 7 - 8 | 1393 | 89 | |
XM_005260938.4 | XP_005260995.1 | 7 - 8 | 1028 | 89 | |
XM_011529520.2 | XP_011527822.1 | 8 - 9 | 1432 | 89 | |
XM_011529521.2 | XP_011527823.1 | 9 - 10 | 1523 | 89 | |
XM_017028313.1 | XP_016883802.1 | 7 - 8 | 1039 | 89 | |
GenBank mRNA | AK223164.1 | - | 7 - 8 | 995 | 89 |
AK313545.1 | - | 7 - 8 | 1190 | 89 | |
BC035031.2 | - | 7 - 8 | 1026 | 89 | |
BX647755.1 | - | 7 - 8 | 1334 | 89 | |
D13318.1 | - | 7 - 8 | 998 | 89 | |
EU159453.1 | - | 6 - 7 | 815 | 89 | |
JQ313900.1 | - | 6 - 7 | 806 | 89 | |
U13044.1 | - | 7 - 8 | 991 | 89 |