The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001326606.1 | NP_001313535.1 | 2 - 3 | 341 | 63 |
NM_001326609.1 | NP_001313538.1 | 3 - 4 | 547 | 63 | |
NM_001326611.1 | NP_001313540.1 | 2 - 3 | 260 | 63 | |
NM_006303.3 | NP_006294.2 | 2 - 3 | 461 | 63 | |
XM_005249847.3 | XP_005249904.1 | 2 - 3 | 675 | 63 | |
XM_005249848.2 | XP_005249905.1 | 2 - 3 | 351 | 63 | |
XM_005249850.3 | XP_005249907.1 | 2 - 3 | 272 | 63 | |
XM_006715772.2 | XP_006715835.1 | 2 - 3 | 246 | 63 | |
XM_011515519.1 | XP_011513821.1 | 2 - 3 | 276 | 63 | |
GenBank mRNA | AK313037.1 | - | 2 - 3 | 467 | 63 |
BC002853.1 | - | 2 - 3 | 421 | 63 | |
BC010156.1 | - | 2 - 3 | 421 | 63 | |
BC013630.1 | - | 2 - 3 | 429 | 63 | |
CX867709.1 | - | 2 - 3 | 313 | 63 | |
U24169.1 | - | 2 - 3 | 461 | 63 |