Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- RAC1
- Entrez Gene ID
-
5879
- Gene Name
- ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1)
- Gene Aliases
-
MIG5, Rac-1, TC-25, p21-Rac1
- Chromosome Location
-
Chr.7: 6374495 - 6403967 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BQ689587.1 |
- |
2 - 3 |
291 |
84 |
View Full Product Details