Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A32
Entrez Gene ID
81034
Gene Name
solute carrier family 25 member 32
Gene Aliases
MFT, MFTC, RREI
Chromosome Location
Chr.8: 103398638 - 103415335 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030780.4 NP_110407.2 3 - 4 791 88
NR_102338.1 4 - 5 900 88
GenBank mRNA AF283645.1 - 3 - 4 521 88
AK027531.1 - 3 - 4 560 88
AK027787.1 - 3 - 4 525 88
AL046268.1 - 3 - 4 361 88
BC021893.1 - 3 - 4 499 88

View Full Product Details