Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DEPDC5
Entrez Gene ID
9681
Gene Name
DEP domain containing 5
Gene Aliases
DEP.5, FFEVF
Chromosome Location
Chr.22: 31753951 - 31907034 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007188.2 NP_001007189.1 10 - 11 826 76
NM_001136029.2 NP_001129501.1 9 - 10 684 76
NM_001242896.1 NP_001229825.1 10 - 11 826 76
NM_001242897.1 NP_001229826.1 10 - 11 730 76
NM_014662.4 NP_055477.1 9 - 10 684 76
NR_110988.1 10 - 11 826 76
XM_005261862.1 XP_005261919.1 10 - 11 730 76
XM_011530557.2 XP_011528859.1 10 - 11 833 76
XM_011530558.2 XP_011528860.1 10 - 11 833 76
XM_011530559.2 XP_011528861.1 10 - 11 833 76
XM_011530560.2 XP_011528862.1 10 - 11 852 76
XM_011530561.2 XP_011528863.1 10 - 11 833 76
XM_011530562.2 XP_011528864.1 10 - 11 833 76
XM_011530563.2 XP_011528865.1 10 - 11 833 76
XM_011530565.2 XP_011528867.1 10 - 11 833 76
XM_011530568.2 XP_011528870.1 10 - 11 833 76
XM_017029109.1 XP_016884598.1 10 - 11 730 76
XM_017029110.1 XP_016884599.1 10 - 11 730 76
XM_017029111.1 XP_016884600.1 10 - 11 730 76
XM_017029112.1 XP_016884601.1 10 - 11 730 76
XM_017029113.1 XP_016884602.1 10 - 11 833 76
XM_017029114.1 XP_016884603.1 10 - 11 833 76
XM_017029115.1 XP_016884604.1 10 - 11 833 76
GenBank mRNA AB014545.1 - 9 - 10 683 76
AJ698950.1 - 9 - 10 679 76
AJ698951.1 - 9 - 10 679 76
AJ704764.1 - 9 - 10 679 76
AK294987.1 - 10 - 11 702 76
AK300737.1 - 9 - 10 711 76
BC057797.1 - 10 - 11 766 76
BC136612.1 - 10 - 11 740 76
BC146766.1 - 9 - 10 683 76

View Full Product Details