Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RBFOX2
- Entrez Gene ID
-
23543
- Gene Name
- RNA binding protein, fox-1 homolog 2
- Gene Aliases
-
FOX2, Fox-2, HNRBP2, HRNBP2, RBM9, RTA, dJ106I20.3, fxh
- Chromosome Location
-
Chr.22: 35738736 - 36028892 on Build GRCh38
View Full Product Details