Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SMTN
Entrez Gene ID
6525
Gene Name
smoothelin
Gene Aliases
-
Chromosome Location
Chr.22: 31081296 - 31104624 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001207017.1 NP_001193946.1 3 - 4 422 117
NM_001207018.1 NP_001193947.1 2 - 3 444 117
NM_006932.4 NP_008863.3 3 - 4 440 117
NM_134269.2 NP_599031.1 3 - 4 440 117
NM_134270.2 NP_599032.2 3 - 4 440 117
XM_006724298.1 XP_006724361.1 3 - 4 418 117
XM_011530333.2 XP_011528635.1 4 - 5 523 117
XM_011530336.2 XP_011528638.1 4 - 5 524 117
XM_011530337.2 XP_011528639.1 4 - 5 524 117
XM_011530338.2 XP_011528640.1 4 - 5 523 117
XM_011530339.2 XP_011528641.1 4 - 5 524 117
XM_011530340.2 XP_011528642.1 4 - 5 524 117
XM_017028913.1 XP_016884402.1 4 - 5 524 117
XM_017028914.1 XP_016884403.1 4 - 5 523 117
XM_017028915.1 XP_016884404.1 4 - 5 523 117
XM_017028916.1 XP_016884405.1 4 - 5 524 117
XM_017028917.1 XP_016884406.1 4 - 5 524 117
XM_017028918.1 XP_016884407.1 4 - 5 524 117
XM_017028919.1 XP_016884408.1 4 - 5 523 117
GenBank mRNA AF064238.3 - 3 - 4 417 117
AJ010306.2 - 3 - 4 417 117
AK128627.1 - 2 - 3 1985 117
AK295418.1 - 2 - 3 444 117
AK300854.1 - 3 - 4 404 117
AK303424.1 - 2 - 3 219 117
AK304084.1 - 3 - 4 422 117
AY061971.1 - 3 - 4 418 117
AY061972.1 - 3 - 4 418 117
BC034237.2 - 3 - 4 408 117
DC342566.1 - 3 - 4 440 117
Y13492.2 - 3 - 4 418 117

View Full Product Details