Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMTN
Entrez Gene ID
6525
Gene Name
smoothelin
Gene Aliases
-
Chromosome Location
Chr.22: 31081296 - 31104624 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134270.2 NP_599032.2 20 - 21 2917 94
GenBank mRNA AY061972.1 - 20 - 21 2895 94

View Full Product Details