Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMTN
Entrez Gene ID
6525
Gene Name
smoothelin
Gene Aliases
-
Chromosome Location
Chr.22: 31081296 - 31104624 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006932.4 NP_008863.3 1 - 2 156 79
NM_134269.2 NP_599031.1 1 - 2 156 79
NM_134270.2 NP_599032.2 1 - 2 156 79
XM_006724298.1 XP_006724361.1 1 - 2 134 79
XM_006724302.1 XP_006724365.1 1 - 2 134 79
GenBank mRNA AK096953.1 - 1 - 2 132 79
AK128719.1 - 1 - 2 134 79
AK293681.1 - 1 - 2 132 79
AK300854.1 - 1 - 2 120 79
BC034237.2 - 1 - 2 124 79
DC342566.1 - 1 - 2 156 79

View Full Product Details