The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135774.1 | NP_001129246.1 | 11 - 12 | 1493 | 138 |
NM_003490.3 | NP_003481.3 | 10 - 11 | 1393 | 138 | |
NM_133633.2 | NP_598344.2 | 10 - 11 | 1393 | 138 | |
XM_011530405.2 | XP_011528707.1 | 11 - 12 | 2029 | 138 | |
XM_011530406.2 | XP_011528708.1 | 11 - 12 | 1495 | 138 | |
XM_011530407.2 | XP_011528709.1 | 11 - 12 | 1902 | 138 | |
XM_011530408.1 | XP_011528710.1 | 11 - 12 | 1493 | 138 | |
XM_011530410.2 | XP_011528712.1 | 9 - 10 | 1697 | 138 | |
XM_017028960.1 | XP_016884449.1 | 12 - 13 | 2427 | 138 | |
XM_017028961.1 | XP_016884450.1 | 11 - 12 | 1819 | 138 | |
XM_017028962.1 | XP_016884451.1 | 11 - 12 | 1754 | 138 | |
XM_017028963.1 | XP_016884452.1 | 11 - 12 | 1418 | 138 | |
XM_017028964.1 | XP_016884453.1 | 11 - 12 | 1490 | 138 | |
XM_017028965.1 | XP_016884454.1 | 5 - 6 | 1264 | 138 | |
GenBank mRNA | AB209684.1 | - | 11 - 12 | 1445 | 138 |
AF046873.1 | - | 10 - 11 | 1270 | 138 | |
AK300171.1 | - | 2 - 3 | 310 | 138 | |
BC075065.2 | - | 10 - 11 | 1260 | 138 | |
BC075066.2 | - | 10 - 11 | 1260 | 138 | |
BC117460.1 | - | 10 - 11 | 1258 | 138 | |
BC143874.1 | - | 10 - 11 | 1261 | 138 | |
CR456589.1 | - | 10 - 11 | 1325 | 138 |