The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018897.2 | NP_061720.2 | 38 - 39 | 6385 | 152 |
XM_011511487.2 | XP_011509789.1 | 39 - 40 | 6761 | 152 | |
XM_011511488.2 | XP_011509790.1 | 37 - 38 | 7435 | 152 | |
XM_011511489.2 | XP_011509791.1 | 37 - 38 | 6347 | 152 | |
XM_011511490.2 | XP_011509792.1 | 37 - 38 | 7330 | 152 | |
XM_011511491.2 | XP_011509793.1 | 38 - 39 | 7557 | 152 | |
XM_011511492.2 | XP_011509794.1 | 38 - 39 | 7557 | 152 | |
XM_011511493.2 | XP_011509795.1 | 38 - 39 | 7557 | 152 | |
XM_011511494.2 | XP_011509796.1 | 38 - 39 | 7556 | 152 | |
XM_011511495.2 | XP_011509797.1 | 38 - 39 | 7556 | 152 | |
XM_011511497.2 | XP_011509799.1 | 5 - 6 | 781 | 152 | |
XM_017004504.1 | XP_016859993.1 | 37 - 38 | 7281 | 152 | |
GenBank mRNA | AB023161.2 | - | 38 - 39 | 6403 | 152 |
AF327442.1 | - | 38 - 39 | 6380 | 152 |