The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005045.3 | NP_005036.2 | 33 - 34 | 5092 | 101 |
NM_173054.2 | NP_774959.1 | 33 - 34 | 5092 | 101 | |
GenBank mRNA | U79716.1 | - | 33 - 34 | 5107 | 101 |